accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHOOCH3OCH3OCH3OONH2CH3CH3OCH3OH3C(E/Z)(E/Z)(E/Z)

CAS RN 70563-58-5

IUPAC Name: 8,13,14,17-tetramethoxy-4,10,12,16-tetramethyl-3,20,22-trioxo-2-azabicyclo[16.3.1]docosa-1(21),4,6,10,18-pentaen-9-yl carbamate
Synonyms: herbimycin
Molecular Weight (g/mol): 574.67
Molecular Formula: C30H42N2O9
InChi Key: MCAHMSDENAOJFZ-UHFFFAOYNA-N
SMILES: COC1CC(C)C(OC)C2=CC(=O)C=C(NC(=O)C(C)=CC=CC(OC)C(OC(N)=O)C(C)=CC(C)C1OC)C2=O
Quantity 
  • (1)
  • (1)
Type 
  • (2)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)