Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Herbimycin A, 95%, Thermo Scientific Chemicals

Catalog No. AAJ66427MCR
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66427MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66427MCR Supplier Thermo Scientific Chemicals Supplier No. J66427MCR
Add to Cart

 

A sparingly soluble benzoquinone anti-tumor agent which interacts with the stress induced chaperone protein Hsp90 (heat shock protein 90)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A sparingly soluble benzoquinone anti-tumor agent which interacts with the stress induced chaperone protein Hsp90 (heat shock protein 90)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70563-58-5
Molecular Formula C30H42N2O9
Molecular Weight (g/mol) 574.671
MDL Number MFCD00151702
InChI Key MCAHMSDENAOJFZ-HYKLGOSOSA-N
PubChem CID 133268477
IUPAC Name [(2S,3R,6S,7R,8E,10R,12E,14E)-2,5,6,11-tetramethoxy-3,7,9,15-tetramethyl-16,20,22-trioxo-17-azabicyclo[16.3.1]docosa-1(21),8,12,14,18-pentaen-10-yl] carbamate
SMILES CC1CC(C(C(C=C(C(C(C=CC=C(C(=O)NC2=CC(=O)C=C(C1OC)C2=O)C)OC)OC(=O)N)C)C)OC)OC

Specifications

Type Herbimycin A
Physical Form Powder
Color Yellow
Quantity 1 mg
Flash Point >110°C (230°F)
Sensitivity Hygroscopic; Light sensitive
Boiling Point 752°C
Beilstein 4834067
Solubility Information Soluble in DMSO
Formula Weight 574.66
Percent Purity 95%
Chemical Name or Material Herbimycin A
Show More Show Less
RTECSNumber LX8930000
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.