accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OFFF

CAS RN 706-27-4

IUPAC Name: 1-methyl-4-(trifluoromethoxy)benzene
Synonyms: 1-methyl-4-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 176.14
Molecular Formula: C8H7F3O
InChi Key: JUXFXYQUXNXVAA-UHFFFAOYSA-N
SMILES: CC1=CC=C(OC(F)(F)F)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results