Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trifluoromethoxy)toluene 98.0+%, TCI America™
SDP

Catalog No. T24545G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24545G 5 g
T24541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24545G Supplier TCI America Supplier No. T24545G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 706-27-4
Molecular Formula C8H7F3O
Molecular Weight (g/mol) 176.138
MDL Number MFCD00042413
InChI Key JUXFXYQUXNXVAA-UHFFFAOYSA-N
Synonym 1-methyl-4-trifluoromethoxy benzene, 4-trifluoromethoxytoluene, 4-trifluoromethoxy toluene, 4-toluene trifluoromethyl ether, 4-methyltrifluoromethoxybenzene, benzene, 1-methyl-4-trifluoromethoxy, 4-methyl-alpha,alpha,alpha-trifluoroanisole, pubchem13486, acmc-209ofc
PubChem CID 2760631
IUPAC Name 1-methyl-4-(trifluoromethoxy)benzene
SMILES CC1=CC=C(C=C1)OC(F)(F)F

Specifications

Color Yellow
Boiling Point 135°C
Quantity 5 g
UN Number 1993
Formula Weight 176.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethoxy)toluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.