accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7063-24-3

IUPAC Name: 6-(4-benzylpiperidin-1-yl)-5-{[(5Z)-3-[2-(3,4-dimethoxyphenyl)ethyl]-4-oxo-2-sulfanylidene-1,3-thiazolidin-5-ylidene]methyl}-1,4-dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile
Synonyms: 6-(4-benzylpiperidin-1-yl)-5-{[(5Z)-3-[2-(3,4-dimethoxyphenyl)ethyl]-4-oxo-2-sulfanylidene-1,3-thiazolidin-5-ylidene]methyl}-1,4-dimethyl-2-oxopyridine-3-carbonitrile
Molecular Weight (g/mol): 628.81
Molecular Formula: C34H36N4O4S2
InChi Key: GYATVUGXRVEMOD-COEJQBHMSA-N
SMILES: COC1=CC=C(CCN2C(=S)S\C(=C/C3=C(N4CCC(CC5=CC=CC=C5)CC4)N(C)C(=O)C(C#N)=C3C)C2=O)C=C1OC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results