Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 4-Nitrophenylacetate 99.0+%, TCI America™
SDP

Catalog No. N039125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N039125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N039125G Supplier TCI America Supplier No. N039125G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7063-24-3
Molecular Formula C34H36N4O4S2
Molecular Weight (g/mol) 628.81
MDL Number MFCD00070537
InChI Key GYATVUGXRVEMOD-COEJQBHMSA-N
Synonym 4-Nitrophenylacetic Acid Sodium Salt
PubChem CID 12675775
IUPAC Name 6-(4-benzylpiperidin-1-yl)-5-{[(5Z)-3-[2-(3,4-dimethoxyphenyl)ethyl]-4-oxo-2-sulfanylidene-1,3-thiazolidin-5-ylidene]methyl}-1,4-dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile
SMILES COC1=CC=C(CCN2C(=S)S\C(=C/C3=C(N4CCC(CC5=CC=CC=C5)CC4)N(C)C(=O)C(C#N)=C3C)C2=O)C=C1OC

Specifications

Color Yellow
Quantity 25 g
Formula Weight 203.13
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 4-Nitrophenylacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.