accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONH(R)H2NOOH

CAS RN 70671-54-4

IUPAC Name: (2R)-6-amino-2-{[(benzyloxy)carbonyl]amino}hexanoic acid
Synonyms: (2R)-6-amino-2-{[(benzyloxy)carbonyl]amino}hexanoic acid| Z-D-LYS-OH
Molecular Weight (g/mol): 280.32
Molecular Formula: C14H20N2O4
InChi Key: OJTJKAUNOLVMDX-GFCCVEGCSA-N
SMILES: NCCCC[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)