Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-Carbobenzoxy-D-lysine 98.0+%, TCI America™
SDP

Catalog No. C21365G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21365G 5 g
C21361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21365G Supplier TCI America Supplier No. C21365G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 70671-54-4 | C14H20N2O4 | 280.32 g/mol

Chemical Identifiers

CAS 70671-54-4
Molecular Formula C14H20N2O4
Molecular Weight (g/mol) 280.32
MDL Number MFCD00067713
InChI Key OJTJKAUNOLVMDX-GFCCVEGCSA-N
Synonym z-d-lys-oh, cbz-d-lysine, nalpha-carbobenzoxy-d-lysine, nalpha-cbz-d-lysine, r-6-amino-2-benzyloxy carbonyl amino hexanoic acid, n-alpha-benzyloxycarbonyl-d-lysine, r-6-amino-2-benzyloxycarbonylamino hexanoic acid, n-cbz-d-lysine, nalpha-z-d-lysine, n, a-cbz-d-lysine
PubChem CID 7017997
IUPAC Name (2R)-6-amino-2-{[(benzyloxy)carbonyl]amino}hexanoic acid
SMILES NCCCC[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 218°C
Color White
Quantity 5 g
Formula Weight 280.32
Percent Purity ≥98.0% (HPLC,T)
Grade Reagent
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-Carbobenzoxy-D-lysine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.