accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 706786-42-7

IUPAC Name: 5-(bromomethyl)-1,3-difluoro-2-methoxybenzene
Synonyms: 5-(bromomethyl)-1,3-difluoro-2-methoxybenzene
Molecular Weight (g/mol): 237.04
Molecular Formula: C8H7BrF2O
InChi Key: IQTBMXJVMUSGSL-UHFFFAOYSA-N
SMILES: COC1=C(F)C=C(CBr)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results