accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 707-07-3

IUPAC Name: (trimethoxymethyl)benzene
Synonyms: trimethoxymethylbenzene
Molecular Weight (g/mol): 182.22
Molecular Formula: C10H14O3
InChi Key: IECKAVQTURBPON-UHFFFAOYSA-N
SMILES: COC(OC)(OC)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (2)
  • (4)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)