accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 707-36-8

IUPAC Name: 1-chloro-3,5-dimethyladamantane
Synonyms: 1-chloro-3,5-dimethyladamantane
Molecular Weight (g/mol): 198.73
Molecular Formula: C12H19Cl
InChi Key: PXDRFQZLDWZHPX-UHFFFAOYNA-N
SMILES: CC12CC3CC(C)(C1)CC(Cl)(C3)C2
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results