Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Chloro-3,5-dimethyladamantane

Catalog No. 802964250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-296-4250MG 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 80-296-4250MG Supplier Sigma Aldrich Supplier No. 802964250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Chloro-3,5-dimethyladamantane

Specifications

CAS 707-36-8
Density 1.0175 g/mL (at 25°C)
Molecular Formula C12H19Cl
Refractive Index n/D 1.4962
Linear Formula C12H19Cl
Quantity 10 mL
MDL Number MFCD00074841
Synonym 1,3-Dimethyl-5-chloroadamantane; 1-chloro-3,5-dimethyl-Tricyclo[3.3.1.13,7 ]decane; 5-Chloro-1,3-dimethyladamantane
Molecular Weight (g/mol) 198.73
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.