accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3SOOBr

CAS RN 707-60-8

IUPAC Name: 4-bromo-N,N-dimethylbenzene-1-sulfonamide
Synonyms: 4-bromo-N,N-dimethylbenzenesulfonamide
Molecular Weight (g/mol): 264.14
Molecular Formula: C8H10BrNO2S
InChi Key: NQAUNPZZVCXYEJ-UHFFFAOYSA-N
SMILES: CN(C)S(=O)(=O)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results