accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 707-99-3

IUPAC Name: 2-(6-amino-9H-purin-9-yl)ethan-1-ol
Synonyms: 2-(6-aminopurin-9-yl)ethanol
Molecular Weight (g/mol): 179.18
Molecular Formula: C7H9N5O
InChi Key: VAQOTZQDXZDBJK-UHFFFAOYSA-N
SMILES: NC1=C2N=CN(CCO)C2=NC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results