Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-(2-Hydroxyethyl)adenine 98.0+%, TCI America™
SDP

Catalog No. H12905G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12905G 5 g
H129025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12905G Supplier TCI America Supplier No. H12905G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 707-99-3
Molecular Formula C7H9N5O
Molecular Weight (g/mol) 179.183
MDL Number MFCD04220770
InChI Key VAQOTZQDXZDBJK-UHFFFAOYSA-N
Synonym 6-Amino-9-(2-hydroxyethyl)purine
PubChem CID 242652
IUPAC Name 2-(6-aminopurin-9-yl)ethanol
SMILES C1=NC2=C(C(=N1)N)N=CN2CCO

Specifications

Melting Point 242°C
Color White-Yellow
Quantity 5 g
Formula Weight 179.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 9-(2-Hydroxyethyl)adenine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.