accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3NNOO

CAS RN 7081-53-0

IUPAC Name: 1-ethyl-4-[2-(morpholin-4-yl)ethyl]-3,3-diphenylpyrrolidin-2-one hydrochloride
Synonyms: doxapram hydrochloride
Molecular Weight (g/mol): 414.97
Molecular Formula: C24H31ClN2O2
InChi Key: MBGXILHMHYLZJT-UHFFFAOYNA-N
SMILES: Cl.CCN1CC(CCN2CCOCC2)C(C1=O)(C1=CC=CC=C1)C1=CC=CC=C1
Color 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)