accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH2N

CAS RN 70849-60-4

IUPAC Name: 4-(2-methylphenyl)piperazin-1-ium
Synonyms: 4-(2-methylphenyl)piperazin-1-ium
Molecular Weight (g/mol): 177.27
Molecular Formula: C11H17N2
InChi Key: WICKLEOONJPMEQ-UHFFFAOYSA-O
SMILES: CC1=CC=CC=C1N1CC[NH2+]CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results