accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COClH3CHOO

CAS RN 7085-19-0

IUPAC Name: 2-(4-chloro-2-methylphenoxy)propanoic acid
Synonyms: mecoprop MCPP mecoprop-P
Molecular Weight (g/mol): 214.65
Molecular Formula: C10H11ClO3
InChi Key: WNTGYJSOUMFZEP-UHFFFAOYNA-N
SMILES: CC(OC1=CC=C(Cl)C=C1C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results