accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3CH3CH3H3C

CAS RN 7087-68-5

IUPAC Name: ethylbis(propan-2-yl)amine
Synonyms: N,N-diisopropylethylamine
Molecular Weight (g/mol): 129.25
Molecular Formula: C8H19N
InChi Key: JGFZNNIVVJXRND-UHFFFAOYSA-N
SMILES: CCN(C(C)C)C(C)C
Quantity 
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (6)
Molecular Weight (g/mol) 
  • (12)
  • (10)
  • (8)
  • (4)
  • (2)
Boiling Point 
  • (2)
  • (3)
  • (3)
  • (9)
  • (4)
  • (12)
  • (2)
Percent Purity 
  • (1)
  • (1)
  • (4)
  • (5)
  • (7)
  • (2)
Physical Form 
  • (20)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (19)
  • (4)
  • (1)
  • (2)
  • (13)
  • (4)
  • (2)
  • (15)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (6)
  • (12)
  • (10)
  • (8)
  • (4)
  • (2)
  • (2)
  • (3)
  • (3)
  • (9)
  • (4)
  • (12)
  • (2)
  • (1)
  • (1)
  • (4)
  • (5)
  • (7)
  • (2)
  • (20)
  • (1)
  • (1)
  • (4)