Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diisopropylethylamine, Biotech Grade, 99.5%,Solstice

Catalog No. 60047000 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
2 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-000 2 L Glass Bottle
60-047-001 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-000 Supplier Solstice Supplier No. 4962192L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotech Grade, 99.5%

  • Solstice developed the first high-purity solvents more than 50 years ago
  • As new applications and techniques emerge, Solstice continues to develop products to meet the most demanding requirements for laboratories and pharmaceutical production
  • Our packaging solutions, ranging from lab scale through production, provide our customers with added value, increased efficiency, and enhanced safety.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7087-68-5
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.247
MDL Number MFCD00008868
InChI Key JGFZNNIVVJXRND-UHFFFAOYSA-N
Synonym n,n-diisopropylethylamine, ethyldiisopropylamine, n-ethyldiisopropylamine, diisopropylethylamine, diea, hunig's base, n-ethyl-n-isopropylpropan-2-amine, dipea, 2-propanamine, n-ethyl-n-1-methylethyl, 1,1'-dimethyltriethylamine
PubChem CID 81531
IUPAC Name N-ethyl-N-propan-2-ylpropan-2-amine
SMILES CCN(C(C)C)C(C)C

Specifications

Melting Point -19.99°C
Density 0.742g/mL at 25°C
Boiling Point 128.33°C
Flash Point 12°C (53.6°F)
Odor Strong, amine like
Packaging Glass Bottle
Linear Formula [(CH3)2CH]2NC2H5
Quantity 2 L
UN Number UN3384
Formula Weight 129.24g/mol
Percent Purity 99.50%
Chemical Name or Material N,N-Diisopropylethylamine
Show More Show Less
Hazard Statement P210-P261-P280-P305 + P351 + P338-P311
DOTInformation 3(6.1)/PGI
RTECSNumber 230-392-0
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.