CAS RN 7089-68-1
IUPAC Name:
2-chloro-1,10-phenanthroline
Synonyms:
2-chloro-1,10-phenanthroline
Molecular Weight (g/mol):
214.65
Molecular Formula:
C12H7ClN2
InChi Key:
JHRMQHFRVPVGHL-UHFFFAOYSA-N
SMILES:
ClC1=CC=C2C=CC3=CC=CN=C3C2=N1
1
–
1
of
1
results
1
2-Chloro-1,10-phenanthroline 98.0+%, TCI America™
CAS: 7089-68-1 Molecular Formula: C12H7ClN2 Molecular Weight (g/mol): 214.65 MDL Number: MFCD00185678 InChI Key: JHRMQHFRVPVGHL-UHFFFAOYSA-N PubChem CID: 355193 IUPAC Name: 2-chloro-1,10-phenanthroline SMILES: ClC1=CC=C2C=CC3=CC=CN=C3C2=N1