Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-1,10-phenanthroline 98.0+%, TCI America™
SDP

Catalog No. C15601G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15601G 1 g
C1560100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15601G Supplier TCI America Supplier No. C15601G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7089-68-1
Molecular Formula C12H7ClN2
Molecular Weight (g/mol) 214.65
MDL Number MFCD00185678
InChI Key JHRMQHFRVPVGHL-UHFFFAOYSA-N
PubChem CID 355193
IUPAC Name 2-chloro-1,10-phenanthroline
SMILES ClC1=CC=C2C=CC3=CC=CN=C3C2=N1

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 1 g
Formula Weight 214.65
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-1,10-phenanthroline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.