accessibility menu, dialog, popup

UserName

Viewing profile as user:
NSOO

CAS RN 709635-68-7

IUPAC Name: 5-(pyridin-2-ylsulfanyl)furan-2-carbaldehyde
Synonyms: 5-(pyridin-2-ylsulfanyl)furan-2-carbaldehyde
Molecular Weight (g/mol): 205.23
Molecular Formula: C10H7NO2S
InChi Key: NWJZBQDKKLHSBP-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(O1)SC1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results