Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(2-Pyridinylsulfanyl)-2-furaldehyde, 95+%, Thermo Scientific™

Catalog No. MO07099DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MO07099DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO07099DA Supplier Thermo Scientific Chemicals Supplier No. MO07099DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 709635-68-7
Molecular Formula C10H7NO2S
Molecular Weight (g/mol) 205.231
InChI Key NWJZBQDKKLHSBP-UHFFFAOYSA-N
Synonym 5-2-pyridinylsulfanyl-2-furaldehyde, 5-pyridin-2-ylthio furan-2-carbaldehyde, 5-pyridin-2-ylsulfanyl furan-2-carbaldehyde, 5-pyridin-2-ylsulfanyl-furan-2-carbaldehyde, 5-pyridin-2-ylthio-2-furaldehyde, 5-2-pyridylthio furan-2-carbaldehyde, 5-2-pyridylsulfanyl furan-2-carbaldehyde, 2-furancarboxaldehyde,5-2-pyridinylthio, 2-furancarboxaldehyde, 5-2-pyridinylthio
PubChem CID 2794784
IUPAC Name 5-pyridin-2-ylsulfanylfuran-2-carbaldehyde
SMILES C1=CC=NC(=C1)SC2=CC=C(O2)C=O

Specifications

Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 205.24
Percent Purity 95%
Chemical Name or Material 5-(2-Pyridinylsulfanyl)-2-furaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.