accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7098-07-9

IUPAC Name: 1-ethyl-1H-imidazole
Synonyms: 1H-imidazole, 1-ethyl-
Molecular Weight (g/mol): 96.13
Molecular Formula: C5H8N2
InChi Key: IWDFHWZHHOSSGR-UHFFFAOYSA-N
SMILES: CCN1C=CN=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results