Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethylimidazole 98.0+%, TCI America™
SDP

Catalog No. E013225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E013225G 25 g
E0132250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E013225G Supplier TCI America Supplier No. E013225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7098-07-9
Molecular Formula C5H8N2
Molecular Weight (g/mol) 96.13
MDL Number MFCD00055389
InChI Key IWDFHWZHHOSSGR-UHFFFAOYSA-N
PubChem CID 81540
IUPAC Name 1-ethylimidazole
SMILES CCN1C=CN=C1

Specifications

Color Yellow
Boiling Point 226°C
Quantity 25 g
Formula Weight 96.13
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Ethylimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.