accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COH

CAS RN 71-41-0

IUPAC Name: pentan-1-ol
Synonyms: amyl alcohol alcool amylique n-C5H11OH pentylalkohol n-amylalkohol amyl alcohol, normal primary amyl alcohol amylalkohol
Molecular Weight (g/mol): 88.15
Molecular Formula: C5H12O
InChi Key: AMQJEAYHLZJPGS-UHFFFAOYSA-N
SMILES: CCCCCO
Molecular Weight (g/mol) 
  • (27)
  • (4)
Quantity 
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (6)
  • (1)
Boiling Point 
  • (13)
  • (3)
  • (4)
  • (6)
Percent Purity 
  • (6)
  • (1)
  • (3)
  • (2)
  • (1)
  • (6)
  • (4)
Grade 
  • (3)
  • (1)
  • (1)
  • (4)
  • (6)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (13)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (6)
  • (2)
  • (2)
  • (10)
  • (1)
  • (27)
  • (4)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (6)
  • (1)
  • (13)
  • (3)
  • (4)
  • (6)
  • (6)
  • (1)
  • (3)
  • (2)
  • (1)
  • (6)
  • (4)
  • (3)
  • (1)
  • (1)
  • (4)
  • (6)
  • (3)
  • (18)
  • (1)
  • (1)