Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Amyl Alcohol, 99.5%, MP Biomedicals

Catalog No. ICN211057 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN211057 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN211057 Supplier MP Biomedicals, Inc Supplier No. 0521105780
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 71-41-0
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00002977,MFCD00081734,MFCD01075169
InChI Key AMQJEAYHLZJPGS-UHFFFAOYSA-N
Synonym 1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol
PubChem CID 6276
ChEBI CHEBI:44884
IUPAC Name pentan-1-ol
SMILES CCCCCO

Specifications

Quantity 100 g
Formula Weight 88.15
Chemical Name or Material n-Amyl Alcohol, 99.5%
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.