accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)(R)(S)(S)CH3(S)(R)OH3COH3CO(R)CH3O

CAS RN 71-58-9

IUPAC Name: (1R,3aS,3bR,5S,9aR,9bS,11aS)-1-acetyl-5,9a,11a-trimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl acetate
Synonyms: provera supprestral methylacetoxyprogesterone preparation strakan brand of medroxyprogesterone repromix repromap depoprovera indivina sirprogen mpa-β
Molecular Weight (g/mol): 386.53
Molecular Formula: C24H34O4
InChi Key: PSGAAPLEWMOORI-PEINSRQWSA-N
SMILES: C[C@H]1C[C@H]2[C@@H]3CC[C@](OC(C)=O)(C(C)=O)[C@@]3(C)CC[C@@H]2[C@@]2(C)CCC(=O)C=C12
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (5)
  • (2)
Grade 
  • (5)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (2)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (2)