Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medroxyprogesterone Acetate, TCI America™
SDP

Catalog No. M19645G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19645G 5 g
M19641G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19645G Supplier TCI America Supplier No. M19645G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 71-58-9
Molecular Formula C24H34O4
Molecular Weight (g/mol) 386.532
MDL Number MFCD00010483
InChI Key PSGAAPLEWMOORI-PEINSRQWSA-N
Synonym medroxyprogesterone acetate, medroxyprogesterone 17-acetate, metigestrona, provera, depo-provera, gestapuran, farlutin, perlutex, veramix, methylacetoxyprogesterone
PubChem CID 6279
ChEBI CHEBI:6716
IUPAC Name [(6S,8R,9S,10R,13S,14S,17R)-17-acetyl-6,10,13-trimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate
SMILES CC1CC2C(CCC3(C2CCC3(C(=O)C)OC(=O)C)C)C4(C1=CC(=O)CC4)C

Specifications

Melting Point 206°C
Color White
Quantity 5 g
Formula Weight 386.53
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Medroxyprogesterone Acetate
EINECSNumber (9)-1470
RTECSNumber TU5010000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.