accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHOFFBr

CAS RN 710291-06-8

IUPAC Name: N-(2-bromophenyl)-2,6-difluorobenzamide
Synonyms: N-(2-bromophenyl)-2,6-difluorobenzamide
Molecular Weight (g/mol): 312.11
Molecular Formula: C13H8BrF2NO
InChi Key: UQJVZAKVKZRKMF-UHFFFAOYSA-N
SMILES: FC1=CC=CC(F)=C1C(=O)NC1=CC=CC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results