accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO

CAS RN 71031-02-2

IUPAC Name: 2-(phenoxymethyl)oxirane
Synonyms: phenyl glydidyl ether
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: FQYUMYWMJTYZTK-UHFFFAOYNA-N
SMILES: C(OC1=CC=CC=C1)C1CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results