Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-Glycidyl Phenyl Ether 95.0+%, TCI America™
SDP

Catalog No. G04091G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G04091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G04091G Supplier TCI America Supplier No. G04091G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 71031-02-2
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD06659025
InChI Key FQYUMYWMJTYZTK-SECBINFHSA-N
Synonym (R)-2-(Phenoxymethyl)oxirane, (R)-1,2-Epoxy-3-phenoxypropane
PubChem CID 1534345
IUPAC Name (2S)-2-(phenoxymethyl)oxirane
SMILES C1C(O1)COC2=CC=CC=C2

Specifications

Color Colorless
Quantity 1 g
Formula Weight 150.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-Glycidyl Phenyl Ether
EINECSNumber (3)-0559&(3)-0594
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.