accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 71042-55-2

IUPAC Name: [3-(diphenylphosphanyl)bicyclo[2.2.1]hept-5-en-2-yl]diphenylphosphane
Synonyms: [3-(diphenylphosphanyl)bicyclo[2.2.1]hept-5-en-2-yl]diphenylphosphane
Molecular Weight (g/mol): 462.51
Molecular Formula: C31H28P2
InChi Key: CDJHPMXMJUCLPA-UHFFFAOYNA-N
SMILES: C1C2C=CC1C(C2P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results