Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2R,3R)-(-)-2,3-Bis(diphenylphosphino)bicyclo[2.2.1]hept-5-ene 98.0+%, TCI America™
SDP

Catalog No. B3449100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3449100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B3449100MG Supplier TCI America Supplier No. B3449100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 71042-55-2
Molecular Formula C31H28P2
Molecular Weight (g/mol) 462.513
MDL Number MFCD00085365
InChI Key CDJHPMXMJUCLPA-ZCVVKFBASA-N
Synonym (2R,3R)-(-)-Norphos
PubChem CID 57853442
IUPAC Name [(2R,3R,4R)-2-diphenylphosphanyl-3-bicyclo[2.2.1]hept-5-enyl]-diphenylphosphane
SMILES C1C2C=CC1C(C2P(C3=CC=CC=C3)C4=CC=CC=C4)P(C5=CC=CC=C5)C6=CC=CC=C6

Specifications

Melting Point 130°C
Color White
Quantity 100 mg
Formula Weight 462.51
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (2R,3R)-(-)-2,3-Bis(diphenylphosphino)bicyclo[2.2.1]hept-5-ene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.