man

Test

Viewing profile as user:
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Main Menu
Admin
Main Menu
ULS Inventory Manager
Products

CAS RN 71058-35-0

CH3CH3NHSNHNH2

CAS RN 71058-35-0

IUPAC Name: 3-amino-1-(2,6-dimethylphenyl)thiourea
Synonyms: 3-amino-1-(2,6-dimethylphenyl)thiourea
Molecular Weight (g/mol): 195.28
Molecular Formula: C9H13N3S
InChi Key: YEOAXWBYZFHOIZ-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=C1NC(=S)NN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results