Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2,6-Dimethylphenyl)-3-thiosemicarbazide, 97%, Thermo Scientific™

Catalog No. AAL1222703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1222703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1222703 Supplier Thermo Scientific Chemicals Supplier No. L1222703

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71058-35-0
Molecular Formula C9H13N3S
Molecular Weight (g/mol) 195.28
MDL Number MFCD00041281
InChI Key YEOAXWBYZFHOIZ-UHFFFAOYSA-N
Synonym 4-2,6-dimethylphenyl-3-thiosemicarbazide, 3-amino-1-2,6-dimethylphenyl thiourea, n-2,6-dimethylphenyl hydrazinecarbothioamide, 1-amino-3-2,6-dimethylphenyl thiourea, 2,6-dimethylphenylthiosemicarbazide, 2,6-dimethylphenyl amino hydrazinomethane-1-thione
PubChem CID 736213
IUPAC Name 3-amino-1-(2,6-dimethylphenyl)thiourea
SMILES CC1=CC=CC(C)=C1NC(=S)NN

Specifications

Melting Point ∽225°C (decomposition)
Quantity 1 g
UN Number UN2811
Beilstein 6768435
Formula Weight 195.29
Percent Purity 97%
Chemical Name or Material 4-(2,6-Dimethylphenyl)-3-thiosemicarbazide
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.