CAS RN 7114-03-6
Thermo Scientific Chemicals Methyl Green, zinc chloride salt, pure, certified
CAS: 7114-03-6 Molecular Formula: C27H35BrClN3·xZnCl2 Molecular Weight (g/mol): 653.23
Thermo Scientific Chemicals Methyl Green, zinc chloride
CAS: 7114-03-6 Molecular Formula: C27H35Cl4N3Zn Molecular Weight (g/mol): 608.78 MDL Number: MFCD00151094 InChI Key: ZKOGXCSOLMZRAZ-UHFFFAOYSA-J Synonym: C.I. 42590 PubChem CID: 131675247 IUPAC Name: zinc;[4-[[4-(dimethylamino)phenyl]-(4-dimethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]phenyl]-ethyl-dimethylazanium;bromide;trichloride SMILES: CC[N+](C)(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-].[Cl-].[Cl-].[Zn+2].[Br-]
Methyl Green, Sigma-Aldrich
A triphenylmethane dye suitable for DNA and RNA detection, used in hematology, histology, and diagnostic assay manufacturing.
Methyl Green, 25g, MP Biomedicals
CAS: 7114-03-6 Molecular Formula: C27H35Cl4N3Zn Molecular Weight (g/mol): 608.78 InChI Key: ZKOGXCSOLMZRAZ-UHFFFAOYSA-J PubChem CID: 131675247 IUPAC Name: zinc;[4-[[4-(dimethylamino)phenyl]-(4-dimethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]phenyl]-ethyl-dimethylazanium;bromide;trichloride SMILES: CC[N+](C)(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-].[Cl-].[Cl-].[Zn+2].[Br-]
Methyl Green (Certified Biological Stain), Fisher Chemical
CAS: 7114-03-6 Molecular Formula: C27H35Cl4N3Zn Molecular Weight (g/mol): 608.78 MDL Number: MFCD00151094 InChI Key: ZKOGXCSOLMZRAZ-UHFFFAOYSA-J Synonym: Ethyl Green PubChem CID: 131675247 IUPAC Name: zinc;[4-[[4-(dimethylamino)phenyl]-(4-dimethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]phenyl]-ethyl-dimethylazanium;bromide;trichloride SMILES: CC[N+](C)(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-].[Cl-].[Cl-].[Zn+2].[Br-]