Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Methyl Green, zinc chloride salt, pure, certified

Catalog No. AC414381000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC414381000 100 g Glass bottle
AC414380250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC414381000 Supplier Thermo Scientific Chemicals Supplier No. 414381000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7114-03-6 | C27H35BrClN3·xZnCl2 | 653.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7114-03-6
Molecular Formula C27H35BrClN3·xZnCl2
Molecular Weight (g/mol) 653.23

Specifications

Melting Point 233.0°C
Color Brown-Red or Green
Physical Form Crystalline Powder
Quantity 100 g
Infrared Spectrum Authentic
Packaging Glass bottle
Merck Index 14,6082
Synonym methanesulfonic anhydride, methanesulfonic acid, anhydride, mesyl anhydride, mesic anhydride, methanesulphonic anhydride, methane sulfonic acid anhydride, methanesulfonyl methanesulfonate, methane sulfonic anhydride, methanesulfonic acid, 1,1'-anhydride, methanesulfonic acid methylsulfonyl ester
Solubility Information Solubility in water: 70g/L (20°C)
InChI Key IZDROVVXIHRYMH-UHFFFAOYSA-N
SMILES CS(=O)(=O)OS(=O)(=O)C
IUPAC Name methylsulfonyl methanesulfonate
PubChem CID 81560
Formula Weight 653.23
Grade Pure
Chemical Name or Material Methyl Green, zinc chloride salt
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 230-415-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.