accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONH(S)NOO

CAS RN 71239-85-5

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(pyridin-2-yl)propanoate
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3-(pyridin-2-yl)propanoate
Molecular Weight (g/mol): 265.29
Molecular Formula: C13H17N2O4
InChi Key: KMODKKCXWFNEIK-JTQLQIEISA-M
SMILES: CC(C)(C)OC(=O)N[C@@H](CC1=CC=CC=N1)C([O-])=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)