Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-3-(2-pyridyl)-L-alanine, 98%

Catalog No. AAH6303706
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6303706 5 g
AAH6303703 1 g
AAH63037MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6303706 Supplier Thermo Scientific Chemicals Supplier No. H6303706
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 71239-85-5 | C13H18N2O4 | 266.297 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71239-85-5
Molecular Formula C13H18N2O4
Molecular Weight (g/mol) 266.297
MDL Number MFCD00191190
InChI Key KMODKKCXWFNEIK-JTQLQIEISA-N
Synonym boc-l-2-pyridylalanine, s-2-tert-butoxycarbonyl amino-3-pyridin-2-yl propanoic acid, boc-3-2-pyridyl-l-alanine, boc-l-3-2-pyridyl-alanine, boc-ala 2-pyridyl-oh, boc-2-pal-oh, boc-3-2-pyridyl-ala-oh, boc-2'-pyridyl-l-ala, n-boc-3-2-pyridyl-l-alanine, boc-l-2-pyridylala
PubChem CID 2734482
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-pyridin-2-ylpropanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=CC=N1)C(=O)O

Specifications

Beilstein 6416850
Quantity 5 g
Formula Weight 266.29
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Boc-3-(2-pyridyl)-L-alanine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.