accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 71255-78-2

IUPAC Name: 2-(2H-1,3-benzodioxol-5-yl)-4,6-bis(trichloromethyl)-1,3,5-triazine
Synonyms: 2-(2H-1,3-benzodioxol-5-yl)-4,6-bis(trichloromethyl)-1,3,5-triazine
Molecular Weight (g/mol): 435.89
Molecular Formula: C12H5Cl6N3O2
InChi Key: BFQFFNWLTHFJOZ-UHFFFAOYSA-N
SMILES: ClC(Cl)(Cl)C1=NC(=NC(=N1)C1=CC2=C(OCO2)C=C1)C(Cl)(Cl)Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results