Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(1,3-Benzodioxol-5-yl)-4,6-bis(trichloromethyl)-1,3,5-triazine 98.0+%, TCI America™
SDP

Catalog No. B36335G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B36335G Supplier TCI America Supplier No. B36335G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 71255-78-2
Molecular Formula C12H5Cl6N3O2
Molecular Weight (g/mol) 435.891
MDL Number MFCD04038074
InChI Key BFQFFNWLTHFJOZ-UHFFFAOYSA-N
PubChem CID 9867644
IUPAC Name 2-(1,3-benzodioxol-5-yl)-4,6-bis(trichloromethyl)-1,3,5-triazine
SMILES C1OC2=C(O1)C=C(C=C2)C3=NC(=NC(=N3)C(Cl)(Cl)Cl)C(Cl)(Cl)Cl

Specifications

Melting Point 135°C
Color Yellow
Quantity 5 g
Formula Weight 435.89
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(1,3-Benzodioxol-5-yl)-4,6-bis(trichloromethyl)-1,3,5-triazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.