accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7128-79-2

IUPAC Name: 3-[(2-methoxyphenyl)carbamoyl]naphthalen-2-yl acetate
Synonyms: 3-[(2-methoxyphenyl)carbamoyl]naphthalen-2-yl acetate
Molecular Weight (g/mol): 335.36
Molecular Formula: C20H17NO4
InChi Key: RKCVQLUEWCVWTN-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1NC(=O)C1=C(OC(C)=O)C=C2C=CC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results