Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyltetrahydroharmol HCl, MP Biomedicals

Catalog No. ICN225289 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN225289 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN225289 Supplier MP Biomedicals, Inc Supplier No. 0522528980

Chemical Identifiers

CAS 7128-79-2
Molecular Formula C20H17NO4
Molecular Weight (g/mol) 335.359
InChI Key RKCVQLUEWCVWTN-UHFFFAOYSA-N
Synonym 2-n-o-methoxyphenyl carbamoyl-3-naphthyl acetate, 3-2-methoxyphenyl carbamoyl naphthalen-2-yl acetate, naphthol-as-ol-acetat, naphthol as-ol acetate, 3-acetoxy-2'-methoxy-2-naphthanilide, 3-n-2-methoxyphenyl carbamoyl-2-naphthyl acetate, 3-acetyloxy-n-2-methoxyphenyl-2-naphthalenecarboxamide, 3-2-methoxyphenyl carbamoyl naphthalen-2-yl ethanoate
PubChem CID 81552
IUPAC Name [3-[(2-methoxyphenyl)carbamoyl]naphthalen-2-yl] acetate
SMILES CC(=O)OC1=CC2=CC=CC=C2C=C1C(=O)NC3=CC=CC=C3OC

Specifications

Quantity 100 mg
Formula Weight 252.7
Chemical Name or Material 2-Methyl Tetrahydroharmol HCL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.