accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 71311-83-6

IUPAC Name: 4-(naphthalen-2-yloxy)aniline
Synonyms: 4-(naphthalen-2-yloxy)aniline
Molecular Weight (g/mol): 235.29
Molecular Formula: C16H13NO
InChi Key: FLSZEMCAUKGBBK-UHFFFAOYSA-N
SMILES: NC1=CC=C(OC2=CC3=CC=CC=C3C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results