Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Aminophenoxy)naphthalene 98.0+%, TCI America™
SDP

Catalog No. A20411G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A20411G Supplier TCI America Supplier No. A20411G

Chemical Identifiers

CAS 71311-83-6
Molecular Formula C16H13NO
Molecular Weight (g/mol) 235.286
MDL Number MFCD00443717
InChI Key FLSZEMCAUKGBBK-UHFFFAOYSA-N
PubChem CID 3580829
IUPAC Name 4-naphthalen-2-yloxyaniline
SMILES C1=CC=C2C=C(C=CC2=C1)OC3=CC=C(C=C3)N

Specifications

Melting Point 116°C
Quantity 1 g
Formula Weight 235.29
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Aminophenoxy)naphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.