accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7147-77-5

ONOOO

CAS RN 7147-77-5

IUPAC Name: 5-(4-nitrophenyl)furan-2-carbaldehyde
Synonyms: 5-(4-nitrophenyl)furan-2-carbaldehyde
Molecular Weight (g/mol): 217.18
Molecular Formula: C11H7NO4
InChi Key: RTSOJVJDKNKNFU-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)C1=CC=C(O1)C=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results