Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(4-Nitrophenyl)-2-furaldehyde 98.0+%, TCI America™
SDP

Catalog No. N08205G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08205G 5 g
N082025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N08205G Supplier TCI America Supplier No. N08205G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7147-77-5
Molecular Formula C11H7NO4
Molecular Weight (g/mol) 217.18
MDL Number MFCD00124191
InChI Key RTSOJVJDKNKNFU-UHFFFAOYSA-N
Synonym 5-4-nitrophenyl-2-furaldehyde, 5-4-nitrophenyl furan-2-carbaldehyde, 5-4-nitrophenyl-2-furancarboxaldehyde, 5-4-nitrophenyl furfural, 5-4-nitro-phenyl-furan-2-carbaldehyde, 2-furancarboxaldehyde, 5-4-nitrophenyl, 5-p-nitrophenyl furfural, 5-4-nitrophenyl 2-furaldehyde, cambridge id 5143178, acmc-1bl92
PubChem CID 81567
IUPAC Name 5-(4-nitrophenyl)furan-2-carbaldehyde
SMILES C1=CC(=CC=C1C2=CC=C(O2)C=O)[N+](=O)[O-]

Specifications

Melting Point 207°C
Quantity 5 g
Formula Weight 217.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-(4-Nitrophenyl)-2-furaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.