accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 71486-53-8

IUPAC Name: hydrogen methyl 4-oxopiperidine-3-carboxylate chloride
Synonyms: hydrogen methyl 4-oxopiperidine-3-carboxylate chloride
Molecular Weight (g/mol): 193.63
Molecular Formula: C7H12ClNO3
InChi Key: NMAACQILAGCQPR-UHFFFAOYNA-N
SMILES: [H+].[Cl-].COC(=O)C1CNCCC1=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)